siRNA Details
| Gene Symbol: Fbxo21 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145564.3 | NP_663539.1 | 2 | 348 | ||
| GenBank mRNA | AB093270.1 | - | 2 | 334 | ||
| AK048131.1 | - | 2 | 327 | |||
| AK048542.1 | - | 2 | 327 | |||
| AK049723.1 | - | 2 | 349 | |||
| AK076521.1 | - | 2 | 345 | |||
| BC021871.1 | - | 2 | 385 | 3709413 | ||

Fbxo21
Genomic Map