siRNA Details
| Gene Symbol: Eva1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145570.2 | NP_663545.2 | 3 | 1259 | ||
| GenBank mRNA | AK145140.1 | - | 3 | 1268 | ||
| BC014699.1 | - | 3 | 1193 | 4221746 | ||
| BC027150.1 | - | 3 | 1215 | 4950341 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Eva1a| Gene Symbol: Eva1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145570.2 | NP_663545.2 | 3 | 1259 | ||
| GenBank mRNA | AK145140.1 | - | 3 | 1268 | ||
| BC014699.1 | - | 3 | 1193 | 4221746 | ||
| BC027150.1 | - | 3 | 1215 | 4950341 | ||