siRNA Details
| Gene Symbol: Zfp428 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001290461.1 | NP_001277390.1 | 3 | 691 | ||
| NM_146183.2 | NP_666295.2 | 3 | 629 | |||
| NM_175142.1 | NP_780351.1 | 3 | 668 | |||
| GenBank mRNA | AK010415.2 | - | 3 | 509 | ||
| AK080517.1 | - | 3 | 622 | |||
| AK139174.1 | - | 3 | 360 | |||
| AV460936.1 | - | 3 | 383 | |||
| BC024802.1 | - | 3 | 530 | 5360536 | ||
| BC096675.1 | - | 3 | 542 | 5146693 | ||
| BX527127.1 | - | 3 | 359 | |||

Zfp428
Genomic Map