siRNA Details
| Gene Symbol: Zfp428 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001290461.1 | NP_001277390.1 | 3 | 863 | ||
| NM_146183.2 | NP_666295.2 | 3 | 801 | |||
| NM_175142.1 | NP_780351.1 | 3 | 840 | |||
| GenBank mRNA | AK010415.2 | - | 3 | 681 | ||
| AK080517.1 | - | 3 | 794 | |||
| AK139174.1 | - | 3 | 532 | |||
| BC024802.1 | - | 3 | 702 | 5360536 | ||
| BC096675.1 | - | 3 | 714 | 5146693 | ||

Zfp428
Genomic Map