siRNA Details
| Gene Symbol: Phldb3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001102613.1 | NP_001096083.1 | 7 | 997 | ||
| GenBank mRNA | BC052555.1 | - | 2 | 301 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Phldb3| Gene Symbol: Phldb3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001102613.1 | NP_001096083.1 | 7 | 997 | ||
| GenBank mRNA | BC052555.1 | - | 2 | 301 | ||