siRNA Details
| Gene Symbol: NPM3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_006993.2 | NP_008924.1 | 4, 5 | 434 | ||
| GenBank mRNA | AF081280.1 | - | 4, 5 | 422 | ||
| AY049737.1 | - | 4, 5 | 426 | |||
| BC041067.1 | - | 4, 5 | 415 | 5473410 | ||
| BC054868.1 | - | 4, 5 | 420 | 6496849 | ||
| BM739536.1 | - | 4, 5 | 434 | |||
| BM765409.1 | - | 4, 5 | 434 | |||

NPM3
Genomic Map