siRNA Details
| Gene Symbol: Zfp420 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172740.2 | NP_766328.2 | 4 | 1274 | ||
| GenBank mRNA | AK045435.1 | - | 2 | 932 | ||
| AK045821.1 | - | 4 | 1187 | |||
| BC050249.1 | - | 3 | 1159 | |||
| BC055817.1 | - | 4 | 1194 | 3595825 | ||

Zfp420
Genomic Map