siRNA Details
| Gene Symbol: Prr12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175022.2 | NP_778187.2 | 4 | 1674 | ||
| GenBank mRNA | AK173130.1 | - | 1 | 1113 | ||
| BC058674.1 | - | 1 | 313 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Prr12| Gene Symbol: Prr12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175022.2 | NP_778187.2 | 4 | 1674 | ||
| GenBank mRNA | AK173130.1 | - | 1 | 1113 | ||
| BC058674.1 | - | 1 | 313 | |||