siRNA Details
| Gene Symbol: Lrrk1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146191.3 | NP_666303.3 | 29 | 4880 | ||
| GenBank mRNA | AK040153.1 | - | 8 | 1385 | ||
| AK076432.1 | - | 2 | 207 | |||
| AK129441.1 | - | 26 | 4293 | |||
| AK147469.1 | - | 29 | 4866 | |||
| AK154395.1 | - | 3 | 399 | |||

Lrrk1
Genomic Map