siRNA Details
| Gene Symbol: BC030336 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001164580.1 | NP_001158052.1 | 2 | 371 | ||
| GenBank mRNA | AK052350.1 | - | 2 | 222 | ||
| AK162324.1 | - | 1 | 64 | |||
| BC030336.1 | - | 2 | 255 | 5401060 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

BC030336| Gene Symbol: BC030336 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001164580.1 | NP_001158052.1 | 2 | 371 | ||
| GenBank mRNA | AK052350.1 | - | 2 | 222 | ||
| AK162324.1 | - | 1 | 64 | |||
| BC030336.1 | - | 2 | 255 | 5401060 | ||