siRNA Details
| Gene Symbol: Klhl26 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001122830.1 | NP_001116302.1 | 2 | 2282 | ||
| NM_178771.3 | NP_848886.1 | 3 | 2465 | |||
| GenBank mRNA | AK036321.1 | - | 3 | 2307 | ||
| AK048601.1 | - | 3 | 2433 | |||
| AK049946.1 | - | 3 | 2465 | |||
| AK163170.1 | - | 3 | 2465 | |||
| AK186605.1 | - | 1 | 97 | |||
| AK218797.1 | - | 1 | 237 | |||
| BC023909.1 | - | 3 | 2480 | 5322205 | ||
| BC025484.1 | - | 3 | 2382 | 5151231 | ||
| BC098185.1 | - | 3 | 2446 | 3594528 | ||

Klhl26
Genomic Map