siRNA Details
| Gene Symbol: Rtbdn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_144929.2 | NP_659178.1 | 6 | 1863 | ||
| GenBank mRNA | AK080715.1 | - | 6 | 1181 | ||
| AY527275.1 | - | 6 | 1863 | |||
| AY527276.1 | - | 7 | 1318 | |||
| BC022744.1 | - | 5 | 1435 | 4527012 | ||
| BC023051.1 | - | 5 | 1391 | 5361063 | ||
| BC027079.1 | - | 5 | 1391 | 5357069 | ||

Rtbdn
Genomic Map