siRNA Details
| Gene Symbol: Rnf214 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_178709.4 | NP_848824.1 | 3 | 980 | ||
| GenBank mRNA | AK028768.1 | - | 3 | 615 | ||
| AK030208.1 | - | 3 | 612 | |||
| AK033867.1 | - | 3 | 476 | |||
| AK046190.1 | - | 3 | 965 | |||
| AK046997.1 | - | 605 | ||||
| AK051527.1 | - | 3 | 981 | |||
| BC094247.1 | - | 2 | 465 | 5717984 | ||

Rnf214
Genomic Map