siRNA Details
| Gene Symbol: Ofd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177429.3 | NP_803178.2 | 2 | 479 | ||
| GenBank mRNA | AJ438159.1 | - | 2 | 330 | ||
| BB652746.1 | - | 2 | 480 | |||
| BC119070.1 | - | 2 | 162 | 8733819 | ||
| BC120487.1 | - | 2 | 162 | 40129410 | ||
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Rat: NM_001106961, XM_008773165, XM_008773162, XM_008773163.

Ofd1
Genomic Map