siRNA Details
| Gene Symbol: Btnl9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172793.2 | NP_766381.1 | 4 | 805 | ||
| GenBank mRNA | AK080918.1 | - | 4 | 789 | ||
| AK084528.1 | - | 4 | 806 | |||
| AY177687.1 | - | 4 | 772 | |||
| BC025540.1 | - | 4 | 783 | |||
| BC052556.1 | - | 3 | 718 | |||
| BC150808.1 | - | 4 | 762 | 9087719 | ||
| BC151155.1 | - | 4 | 762 | 9088043 | ||

Btnl9
Genomic Map