siRNA Details
| Gene Symbol: Zdhhc22 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001080943.2 | NP_001074412.1 | 2 | 697 | ||
| GenBank mRNA | AK080584.1 | - | 1 | 361 | ||
| BC128021.1 | - | 2 | 701 | 40136338 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Zdhhc22| Gene Symbol: Zdhhc22 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001080943.2 | NP_001074412.1 | 2 | 697 | ||
| GenBank mRNA | AK080584.1 | - | 1 | 361 | ||
| BC128021.1 | - | 2 | 701 | 40136338 | ||