siRNA Details
| Gene Symbol: KLF2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_016270.3 | NP_057354.1 | 3 | 1497 | ||
| GenBank mRNA | AF134053.1 | - | 3 | 1399 | ||
| AF205849.1 | - | 3 | 1483 | |||
| AK225820.1 | - | 3 | 1496 | |||
| BC071983.1 | - | 3 | 1486 | 5480246 | ||
| BQ581981.1 | - | 2 | 533 | |||

KLF2
Genomic Map