siRNA Details
| Gene Symbol: Zfp760 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001008501.2 | NP_001008501.2 | 6 | 1419 | ||
| GenBank mRNA | BC085182.1 | - | 6 | 1419 | 30653533 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Zfp760| Gene Symbol: Zfp760 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001008501.2 | NP_001008501.2 | 6 | 1419 | ||
| GenBank mRNA | BC085182.1 | - | 6 | 1419 | 30653533 | |