siRNA Details
| Gene Symbol: Dmxl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081371.2 | NP_001074840.2 | 24 | 5950 | ||
| GenBank mRNA | AY590892.1 | - | 24 | 5950 | ||
| BC056490.1 | - | 1 | 114 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Dmxl1| Gene Symbol: Dmxl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081371.2 | NP_001074840.2 | 24 | 5950 | ||
| GenBank mRNA | AY590892.1 | - | 24 | 5950 | ||
| BC056490.1 | - | 1 | 114 | |||