siRNA Details
| Gene Symbol: Lrrc7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081358.2 | NP_001074827.1 | 5, 6 | 1603 | ||
| NM_001291452.1 | NP_001278381.1 | 5, 6 | 1603 | |||
| NM_001291453.1 | NP_001278382.1 | 5, 6 | 1603 | |||
| GenBank mRNA | AK122498.1 | - | 5, 6 | 738 | ||
| AK141618.1 | - | 5, 6 | 1603 | |||
| BC138450.1 | - | 5, 6 | 1510 | 8861470 | ||

Lrrc7
Genomic Map