siRNA Details
| Gene Symbol: Tmem245 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175518.5 | NP_780727.3 | 14 | 2106 | ||
| GenBank mRNA | AK052826.1 | - | 14 | 2665 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tmem245| Gene Symbol: Tmem245 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175518.5 | NP_780727.3 | 14 | 2106 | ||
| GenBank mRNA | AK052826.1 | - | 14 | 2665 | ||