siRNA Details
| Gene Symbol: Rab42 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081651.1 | NP_001075120.1 | 2 | 417 | ||
| GenBank mRNA | AB232642.1 | - | 2 | 398 | ||
| AK035722.1 | - | 1 | 761 | |||
| BC132060.1 | - | 2 | 446 | 40130337 | ||
| BC132062.1 | - | 2 | 446 | 40130339 | ||

Rab42
Genomic Map