siRNA Details
| Gene Symbol: Tshz3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172298.2 | NP_758502.1 | 2 | 3296 | ||
| GenBank mRNA | AK037176.1 | - | 1 | 619 | ||
| AK041793.1 | - | 1 | 2182 | |||
| AK086984.1 | - | 1 | 1679 | |||
| AK220396.1 | - | 2 | 3167 | |||
| AY063491.1 | - | 2 | 3296 | |||
| BC150674.1 | - | 2 | 3239 | 9087585 | ||

Tshz3
Genomic Map