siRNA Details
| Gene Symbol: Rgma | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177740.5 | NP_808408.2 | 4 | 1562 | ||
| GenBank mRNA | AJ557513.1 | - | 3 | 1316 | ||
| AK082560.1 | - | 3 | 1459 | |||
| AK145224.1 | - | 3 | 1307 | |||
| BC023870.1 | - | 3 | 1370 | 5353958 | ||
| BC059072.1 | - | 4 | 1562 | 6826181 | ||

Rgma
Genomic Map