siRNA Details
| Gene Symbol: Zc4h2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001003916.2 | NP_001003916.1 | 2 | 290 | ||
| NM_001289696.1 | NP_001276625.1 | 2 | 290 | |||
| NM_001289697.1 | NP_001276626.1 | 2 | 290 | |||
| GenBank mRNA | AK129302.1 | - | 2 | 164 | ||
| AK155445.1 | - | 2 | 290 | |||
| AK162005.1 | - | 2 | 187 | |||
| BC050850.1 | - | 2 | 188 | 6508566 | ||
| BC079862.1 | - | 2 | 213 | 6807437 | ||
| BC094942.1 | - | 2 | 231 | 6435125 | ||
| BC119285.1 | - | 2 | 204 | 8734034 | ||
| BC119311.1 | - | 2 | 205 | 8734060 | ||
| BY200912.1 | - | 2 | 290 | |||

Zc4h2
Genomic Map