siRNA Details
| Gene Symbol: Olfr370 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC119367.1 | - | 1 | 696 | 8734116 | |
| BC120734.1 | - | 1 | 696 | 40129657 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr370| Gene Symbol: Olfr370 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC119367.1 | - | 1 | 696 | 8734116 | |
| BC120734.1 | - | 1 | 696 | 40129657 | ||