siRNA Details
| Gene Symbol: Olfr742 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146430.2 | NP_666641.2 | 1 | 191 | ||
| GenBank mRNA | BC132583.1 | - | 1 | 191 | 40130860 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr742| Gene Symbol: Olfr742 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146430.2 | NP_666641.2 | 1 | 191 | ||
| GenBank mRNA | BC132583.1 | - | 1 | 191 | 40130860 | |
Set Membership: |
|