siRNA Details
| Gene Symbol: Olfr360 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146622.1 | NP_666833.1 | 1 | 576 | ||
| GenBank mRNA | BC110546.2 | - | 1 | 574 | ||
| BC125238.1 | - | 1 | 574 | |||
| BC125239.1 | - | 1 | 575 | |||
| BC126918.1 | - | 1 | 575 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr360| Gene Symbol: Olfr360 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146622.1 | NP_666833.1 | 1 | 576 | ||
| GenBank mRNA | BC110546.2 | - | 1 | 574 | ||
| BC125238.1 | - | 1 | 574 | |||
| BC125239.1 | - | 1 | 575 | |||
| BC126918.1 | - | 1 | 575 | |||