siRNA Details
Gene Symbol: Olfr739 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146668.2 | NP_666879.2 | 1 | 602 |

More Information
Set Membership: |
![]() |
Panther Classification:
Gene Ontology Categories:
