siRNA Details
| Gene Symbol: Olfr926 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146815.1 | NP_667026.1 | 1 | 757 | ||
| GenBank mRNA | BC120558.1 | - | 1 | 809 | 40129481 | |
| BC125490.1 | - | 1 | 809 | 40130005 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr926| Gene Symbol: Olfr926 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146815.1 | NP_667026.1 | 1 | 757 | ||
| GenBank mRNA | BC120558.1 | - | 1 | 809 | 40129481 | |
| BC125490.1 | - | 1 | 809 | 40130005 | ||