siRNA Details
| Gene Symbol: Olfr376 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001172686.1 | NP_001166157.1 | 3 | 916 | ||
| NM_146922.2 | NP_667133.1 | 2 | 785 | |||
| Gene Symbol: Olfr20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC138172.1 | - | 1 | 671 | 8861191 | |
| BC138173.1 | - | 1 | 671 | 8861192 | ||

Olfr376
Genomic Map