siRNA Details
| Gene Symbol: Olfr1413 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_147037.2 | NP_667248.1 | 1 | 734 | ||
| GenBank mRNA | BC116898.1 | - | 1 | 734 | 40126217 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr1413| Gene Symbol: Olfr1413 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_147037.2 | NP_667248.1 | 1 | 734 | ||
| GenBank mRNA | BC116898.1 | - | 1 | 734 | 40126217 | |