siRNA Details
| Gene Symbol: Olfr686 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_147069.1 | NP_667280.1 | 1 | 473 | ||
| GenBank mRNA | BC132004.1 | - | 1 | 508 | 40130281 | |
| BC132006.1 | - | 1 | 508 | 40130283 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr686| Gene Symbol: Olfr686 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_147069.1 | NP_667280.1 | 1 | 473 | ||
| GenBank mRNA | BC132004.1 | - | 1 | 508 | 40130281 | |
| BC132006.1 | - | 1 | 508 | 40130283 | ||