siRNA Details
| Gene Symbol: Olfr550 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_147104.2 | NP_667315.2 | 1 | 687 | ||
| GenBank mRNA | BC116960.1 | - | 1 | 687 | 40126279 | |
| BC116964.1 | - | 1 | 687 | 40126283 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr550| Gene Symbol: Olfr550 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_147104.2 | NP_667315.2 | 1 | 687 | ||
| GenBank mRNA | BC116960.1 | - | 1 | 687 | 40126279 | |
| BC116964.1 | - | 1 | 687 | 40126283 | ||