siRNA Details
| Gene Symbol: Esrrb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001159500.1 | NP_001152972.1 | 6 | 1282 | ||
| NM_011934.4 | NP_036064.3 | 6 | 1181 | |||
| GenBank mRNA | AK032025.1 | - | 6 | 989 | ||
| AK044339.1 | - | 6 | 1182 | |||
| AK052256.1 | - | 6 | 1284 | |||
| BC044858.1 | - | 5 | 851 | |||
| BC132595.1 | - | 6 | 1146 | 40130872 | ||
| BC132597.1 | - | 6 | 1147 | 40130874 | ||
| S82458.1 | - | 5 | 889 | |||
| X89594.1 | - | 5 | 889 | |||

Esrrb
Genomic Map