siRNA Details
| Gene Symbol: Ube2z | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172300.3 | NP_758504.3 | 7 | 1055 | ||
| GenBank mRNA | AK005524.1 | - | 5 | 606 | ||
| AK077793.1 | - | 7 | 1054 | |||
| AK149770.1 | - | 7 | 1055 | |||
| AK152529.1 | - | 7 | 769 | |||
| AK153328.1 | - | 7 | 786 | |||
| AK168942.1 | - | 7 | 748 | |||
| AK180312.1 | - | 1 | 47 | |||
| AK187506.1 | - | 1 | 47 | |||
| BC027266.1 | - | 7 | 680 | |||
| BC054412.1 | - | 7 | 758 | 2646768 | ||

Ube2z
Genomic Map