siRNA Details
| Gene Symbol: Cenph | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021886.1 | NP_068686.1 | 6 | 477 | ||
| GenBank mRNA | AB017634.1 | - | 6 | 477 | ||
| AK006208.1 | - | 6 | 461 | |||
| AK010533.1 | - | 3 | 539 | |||
| AK011264.1 | - | 6 | 462 | |||
| AK011738.1 | - | 6 | 462 | |||
| AK012907.1 | - | 6 | 422 | |||
| BC025084.1 | - | 6 | 428 | 3501244 | ||

Cenph
Genomic Map