siRNA Details
| Gene Symbol: Krtap13-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_183189.1 | NP_899012.1 | 1 | 360 | ||
| GenBank mRNA | BC027551.1 | - | 1 | 360 | 1363815 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Krtap13-1| Gene Symbol: Krtap13-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_183189.1 | NP_899012.1 | 1 | 360 | ||
| GenBank mRNA | BC027551.1 | - | 1 | 360 | 1363815 | |