siRNA Details
| Gene Symbol: Ccdc32 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_199310.2 | NP_955014.2 | 4 | 986 | ||
| GenBank mRNA | AK132604.1 | - | 4 | 978 | ||
| AK133813.1 | - | 4 | 961 | |||
| AK147152.1 | - | 4 | 962 | |||
| AK152091.1 | - | 1 | 339 | |||
| BC023926.1 | - | 4 | 980 | 5324154 | ||
| BC052687.1 | - | 4 | 953 | |||

Ccdc32
Genomic Map