siRNA Details
| Gene Symbol: Gpr157 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177366.3 | NP_796340.2 | 3 | 1032 | ||
| GenBank mRNA | AK089429.1 | - | 3 | 1172 | ||
| AK089544.1 | - | 3 | 985 | |||
| AK154291.1 | - | 3 | 668 | |||
| AK170941.1 | - | 3 | 627 | |||
| AK171081.1 | - | 3 | 627 | |||
| BC117996.1 | - | 3 | 986 | 40095824 | ||
More Information
Set Membership: |
|

Gpr157
Genomic Map