siRNA Details
Gene Symbol: Fat3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080814.1 | NP_001074283.1 | 20 | 11564 | ||
GenBank mRNA | AK035028.1 | - | 3 | 369 | ||
BC117744.1 | - | 3 | 344 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Fat3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080814.1 | NP_001074283.1 | 20 | 11564 | ||
GenBank mRNA | AK035028.1 | - | 3 | 369 | ||
BC117744.1 | - | 3 | 344 |