siRNA Details
| Gene Symbol: Nlrx1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001163742.1 | NP_001157214.1 | 7 | 1926 | ||
| NM_001163743.1 | NP_001157215.1 | 7 | 2027 | |||
| NM_178420.3 | NP_848507.2 | 7 | 2015 | |||
| GenBank mRNA | AK141296.1 | - | 7 | 1997 | ||
| AK166689.1 | - | 7 | 1853 | |||
| AK170088.1 | - | 7 | 1970 | |||
| BC034204.1 | - | 6 | 1809 | |||
| BC035045.1 | - | 1 | 140 | |||
| BC050054.1 | - | 7 | 1922 | 30012053 | ||

Nlrx1
Genomic Map