siRNA Details
| Gene Symbol: Klhdc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_178253.5 | NP_839984.1 | 4 | 473 | ||
| GenBank mRNA | AK144621.1 | - | 4 | 413 | ||
| AY238938.1 | - | 4 | 473 | |||
| BC119567.2 | - | 4 | 457 | 40057793 | ||
| BC119568.2 | - | 4 | 457 | 40057794 | ||

Klhdc1
Genomic Map