siRNA Details
| Gene Symbol: SCML2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_006089.2 | NP_006080.1 | 7 | 740 | ||
| NR_033717.1 | 7 | 740 | ||||
| XM_006724458.1 | XP_006724521.1 | 7 | 704 | |||
| XM_017029219.1 | XP_016884708.1 | 7 | 636 | |||
| XM_017029220.1 | XP_016884709.1 | 7 | 632 | |||
| XM_017029221.1 | XP_016884710.1 | 7 | 774 | |||
| GenBank mRNA | AK303064.1 | - | 6 | 606 | ||
| AK315116.1 | - | 7 | 704 | |||
| BC051913.1 | - | 7 | 682 | |||
| BC064617.1 | - | 7 | 697 | 5575894 | ||
| Y18004.1 | - | 7 | 674 | |||

SCML2
Genomic Map