siRNA Details
| Gene Symbol: Snf8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_033568.2 | NP_291046.1 | 4 | 405 | ||
| GenBank mRNA | AK013058.1 | - | 4 | 407 | ||
| AK020203.1 | - | 2 | 95 | |||
| BC003938.1 | - | 4 | 377 | 3486703 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Snf8| Gene Symbol: Snf8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_033568.2 | NP_291046.1 | 4 | 405 | ||
| GenBank mRNA | AK013058.1 | - | 4 | 407 | ||
| AK020203.1 | - | 2 | 95 | |||
| BC003938.1 | - | 4 | 377 | 3486703 | ||