siRNA Details
| Gene Symbol: BMP5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021073.2 | NP_066551.1 | 6 | 1910 | ||
| XM_017011198.1 | XP_016866687.1 | 5 | 615 | |||
| GenBank mRNA | AK291559.1 | - | 6 | 1923 | ||
| BC027958.1 | - | 6 | 1910 | 5209637 | ||
| KC485577.1 | - | 6 | 1198 | |||
| M60314.1 | - | 6 | 1896 | |||

BMP5
Genomic Map