siRNA Details
| Gene Symbol: Kiss1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_178260.3 | NP_839991.2 | 1 | 81 | ||
| GenBank mRNA | AA023297.1 | - | 1 | 64 | ||
| AB162440.1 | - | 1 | 43 | |||
| AB666166.1 | - | 2 | 241 | |||
| AF472576.1 | - | 1 | 81 | |||
| BC117046.1 | - | 1 | 66 | 40126365 | ||
| BC119348.1 | - | 1 | 66 | 8734097 | ||

Kiss1
Genomic Map