siRNA Details
| Gene Symbol: Rilp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001029938.2 | NP_001025109.1 | 7 | 1005 | ||
| GenBank mRNA | AK139016.1 | - | 7 | 997 | ||
| AK160517.1 | - | 5 | 798 | |||
| BC038146.1 | - | 3 | 323 | |||
| BC051502.1 | - | 4 | 464 | |||
| BC116750.1 | - | 7 | 989 | 40126069 | ||
| BC119166.1 | - | 7 | 989 | 8733915 | ||

Rilp
Genomic Map