siRNA Details
| Gene Symbol: Cep63 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081122.1 | NP_001074591.1 | 11 | 1252 | ||
| NM_001301689.1 | NP_001288618.1 | 11 | 1252 | |||
| GenBank mRNA | AK076549.1 | - | 11 | 1253 | ||
| BC048718.1 | - | 4 | 404 | |||
| KC211185.1 | - | 11 | 1109 | |||

Cep63
Genomic Map