siRNA Details
| Gene Symbol: Mga | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001164274.1 | NP_001157746.1 | 2 | 333 | ||
| NM_013720.2 | NP_038748.2 | 2 | 333 | |||
| GenBank mRNA | AF205935.1 | - | 1 | 147 | ||
| AK012050.1 | - | 2 | 296 | |||
| AK032570.1 | - | 2 | 314 | |||
| AK046252.1 | - | 2 | 334 | |||
| AK133775.1 | - | 2 | 550 | |||
| AK139919.1 | - | 2 | 285 | |||
| AK145491.1 | - | 2 | 282 | |||
| AK162302.1 | - | 2 | 330 | |||
| AK172962.1 | - | 2 | 256 | |||

Mga
Genomic Map